Would you like to log in again?
Proto® Prick Punches, ORS Nasco
Supplier: ORS Nasco
577-443/8 577-445/16
76188-878EA 9.44 USD InStock
76188-878 76188-870
Proto® Prick Punches, ORS Nasco
Chisels
Aligns holes in two or more pieces of metal through which a bolt or screw is to be inserted.
  • Tool steel
  • Round tip
  • Hex stock shape
Modified parabolic striking end for greater resistance to mushrooming and chipping. For sheet metal working layout such as scribing and for punch starting holes for sheet metal screws.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty