Would you like to log in again?
Proto® 12-Point Self-Aligning Ratcheting Box Wrenches, ORS Nasco
Supplier: ORS Nasco
577-1187M-A 577-1186M-A 577-1185M-A
76190-424EA 46.49 USD InStock
76190-424 76190-428 76190-420
Proto® 12-Point Self-Aligning Ratcheting Box Wrenches, ORS Nasco
Wrenches
Fits 4, 6 and 12 point fasteners.
  • Box/box opening
  • Metric
  • Off-set handle
  • Full polish finish
Full polish finish provides corrosion resistance and easy to clean surfaces. 12 point box end allows for fine indexing. Built in retaining ring in each ratchet wheel adds speed and convenience. Reversing switch lever allows for easy changes in turning direction which helps improve speed. Shorter length for access to confined workspaces. The wrench self-aligns with the fastener.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty