Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Proto® Heavy-Duty Offset Striking Wrenches, ORS Nasco
Supplier: ORS Nasco
577-2643SW 577-2619SW 577-2622SW 577-2660SW 577-2625SW 577-2662SW 577-2631SW 577-2654SW 577-2656SW 577-2639SW 577-2646SW 577-2616SW 577-2668SW 577-2666SW
76188-724EA 690.54 USD InStock
76188-724 76188-702 76188-734 76188-698 76188-720 76188-644 76188-732 76188-664 76188-692 76190-634 76190-642 76190-654 76188-728 76188-716
Proto® Heavy-Duty Offset Striking Wrenches, ORS Nasco
Wrenches
Large striking surface.
  • Black oxide finish
  • Off-set handle
  • Box opening
  • Inch measuring system
  • Forged alloy steel
Heavy box walls provide greater torque transmission. Short handle permits work in close quarters. Offset provides clearance for knuckles and obstructions. Box opening remains on fastener when wrench is struck. Efficient transmittal of force to fastener. For where extreme force is needed to loosen or set large fasteners.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty