Would you like to log in again?
Proto® Micrometer Ratcheting Head Torque Wrenches, ORS Nasco
Supplier: ORS Nasco
577-6014MC 577-6006MC
76189-696EA 434.5 USD InStock
76189-696 76189-706
Proto® Micrometer Ratcheting Head Torque Wrenches, ORS Nasco
Wrenches
Calibration is accomplished by manual loading the torque wrenches.
  • Micrometer torque setting
  • Ball locking
  • Pear head
  • Male square drive
  • Alloy steel
Manufactured to meet standard industrial torque applications, general industry, heavy equipment, and maintenance. Calibrated to +/- 3% clockwise direction and +/- 6% counter clockwise direction at 20% to 100% of full scale.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty