Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Proto® Retaining Rings, Essendant
Supplier: Stanley
577-RR50115 577-RR07520 577-RR07534 577-RR50160 577-RR07532 577-RR50145 577-RR50105 577-RR10042 577-RR07528 577-RR07536 577-RR50087 577-RR50080 577-RR50130 577-RR50095
76191-268EA 113.56 USD InStock
76191-268 76190-060 76190-050 76190-054 76190-052 76190-046 76190-058 76190-044 76190-056 76190-048 76191-270 76191-262 76191-264 76191-266
Proto® Retaining Rings, Essendant
Wrenches
Helps secure sockets onto drive tools.
  • One-piece construction, no loose parts
  • O-ring type
Resin crush gauge on the pin helps indicate when a socket is excessively worn and should be replaced.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty