Would you like to log in again?
UltraBrite™ Yellow IHC Chromogen (HRP), BioVision
Supplier: BioVision
AB286894-30ML
76182-476EA 1179.43 USD InStock
76182-476
UltraBrite™ Yellow IHC Chromogen (HRP), BioVision
Stains Microbiology Stains
Unique yellow IHC chromogen for HRP-activated development.
  • APPLICATIONS: IHC, ISH (in situ hybridization)
The UltraBrite™ yellow IHC chromogen (HRP) is a substrate chromogen system designed to be used for either immunohistochemistry (IHC) or ISH when utilizing horseradish peroxidase. UltraBrite™ yellow IHC chromogen (HRP) produces a distinct bright yellow that can easily be distinguished from other stains.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty