Would you like to log in again?
Tiger Disc Angled Style Flap Discs, Weiler
Supplier: Weiler
804-50510 804-50565
76183-928EA 173.54 USD InStock
76183-928 76183-930
Tiger Disc Angled Style Flap Discs, Weiler
Abrasives
Use for grinding, finishing, weld blending, and deburring applications
  • Maximum aggression and long life
  • Ideal for a working angle of 15° to 25°
Ideal for use on steel, most metals, cast iron, and non-ferrous soft metal materials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty