Would you like to log in again?
Saber Tooth Ceramic Resin Fiber Discs, Weiler®
Supplier: Weiler
804-59560 804-59551 804-59566
76184-308EA 5.17 USD InStock
76184-308 76184-306 76184-304
Saber Tooth Ceramic Resin Fiber Discs, Weiler®
Abrasives
Designed for all-purpose weld blending, grinding, surface preparation, and finishing
Use with a right angle grinder on aluminum, stainless steel, and other hard-to-grind metals.
Resin bond technology with superior grain adhesion offers extended life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty