Would you like to log in again?
Weiler® Copper Center™ Small Diameter Wire Wheel, ORS Nasco
Supplier: ORS Nasco
804-15141
76183-792CT 113.54 USD InStock
76183-792
Weiler® Copper Center™ Small Diameter Wire Wheel, ORS Nasco
Brushes
Use with die grinders, right-angle air die grinders, or power drills to remove rust, paint, oxides, and scale.
  • Wire fill
  • Steel filter material
  • Can be mounted in multiples
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty