Would you like to log in again?
Knot Wire Bevel Wheel, Weiler®
Supplier: Weiler
804-13456
76183-788EA 45.48 USD InStock
76183-788
Knot Wire Bevel Wheel, Weiler®
Abrasives
Maximum aggression for cleaning in corners.
  • Heavy-duty brushing action with some flexibility
  • Ideal for cleaning filet welds
  • Steel filter material
Use with right-angled grinders to remove rust, paint, oxides, and scale.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty