Would you like to log in again?
Medium Sweeping Broom Display Packs, Weiler®, ORS Nasco
Supplier: ORS Nasco
804-36634 804-36636
76183-924EA 482.05 USD InStock
76183-924 76183-926
Medium Sweeping Broom Display Packs, Weiler®, ORS Nasco
Brooms
Delivers true flexibility when switching between cutting and grinding for stainless steel applications.
  • Contaminate-free white aluminum oxide wheel
  • Fiberglass body
  • Resin bonded
  • Long lasting and fast cutting
  • Use with right angle grinders
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty