Would you like to log in again?
Miniature Stem-Mounted Wheel Brush, Weiler
Supplier: Weiler
804-26003
76183-822EA 774.72 USD InStock
76183-822
Miniature Stem-Mounted Wheel Brush, Weiler
Abrasives
Miniature stem-mounted wheel brushes are a great choice for cleaning and deburring with a flexible shaft or straight grinder.
  • 0.005" steel bristle
  • For use with high-speed electric rotary tools or air-powered pencil die grinders
Use them for buffing and polishing of small parts. Also good for decorative finishing and edge blending.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty