Would you like to log in again?
Narrow Face Crimped Wire Wheel, Weiler®
Supplier: Weiler
804-01250-12 804-01068 804-01695
76183-712EA 112.69 USD InStock
76183-712 76183-796 76183-798
Narrow Face Crimped Wire Wheel, Weiler®
Abrasives
Use to remove rust, light corrosion, paint, oxides, scale, or heat discoloration
  • 0.014" steel bristle
  • Flexible brushing action
  • Can be mounted in multiples
  • Consistent performance
Compatible with straight grinders, bench/pedestal grinders, and right angle grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty