Would you like to log in again?
Wolverine Thin Cutting Wheels, Weiler
Supplier: Weiler
804-56281
76184-170EA 2.72 USD InStock
76184-170
Wolverine Thin Cutting Wheels, Weiler
Abrasives
Resin-bonded disc is formulated for aggressive stock removal and fast cutting
  • Reinforced with fiberglass webbing for stability
  • Use with a thin cutting wheel, cut-off die grinder, or right angle air die grinder
  • Use for cutting and parting, notching, rough grinding, and weld removal applications
Suitable for general purpose cutting and grinding applications on steel, iron, and other ferrous metals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty