Would you like to log in again?
Wide Face Crimped Wire Wheel, Weiler
Supplier: Weiler
804-03210
76183-714EA 300.37 USD InStock
76183-714
Wide Face Crimped Wire Wheel, Weiler
Abrasives
Maximum wire density and aggression, covers more surface area in a single pass
  • Provides flexible brushing action
  • Use with bench/pedastal grinders
  • Steel filter material
Removes rust, paint, oxides, and scale.
Consistent and effective performance for cleaning, light deburring, edge blending, roughening, and finishing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty