Would you like to log in again?
Double Row Heavy-Duty Knot Wire Cup Brush, Weiler®
Supplier: Weiler
804-12576
76183-778EA 170.61 USD InStock
76183-778
Double Row Heavy-Duty Knot Wire Cup Brush, Weiler®
Abrasives
Knot wire cup brush provides very aggressive brushing action while still maintaining some flexibility
  • Ideal tool for severe cleaning applications
  • Use with right-angled grinders
  • 0.035" steel bristle
Use with right-angled grinders to remove rust, paint, oxides, and scale.
Best suited for demanding cleaning applications on large surface areas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty