Would you like to log in again?
Tiger, Zirconia Ultracut Thin Cutting Wheel, Weiler
Supplier: Weiler
Tiger®
804-58005 804-58006
76184-204EA 219.9 USD InStock
76184-204 76184-206
Tiger, Zirconia Ultracut Thin Cutting Wheel, Weiler
Abrasives
Self-sharpening Zirconia Alumina grain for excellent performance on thin gauge and small diameter steel and stainless steel applications.
  • Delivers precise, clean, 1mm thick cuts
  • Features Solid Core Technology for reduced friction and longer life
  • Zirconia Alumina cutting edge
  • Z60T grind
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty