Would you like to log in again?
Tiger, Ceramic Grinding Wheels, Weiler®
Supplier: Weiler
Tiger®
804-58331 804-58332 804-58325 804-58326
76184-248BX 153.56 USD InStock
76184-248 76184-250 76184-262 76184-260
Tiger, Ceramic Grinding Wheels, Weiler®
Abrasives
Self-sharpening ceramic alumina grain delivers a superior combination of life and cut rate
  • Superior life and cut on metal
Use for cutting, edge beveling, edge softening, grinding, notching, scale removal, stock removal, weld blending, and weld removal applications.
Ideal for stainless steel and other hard-to-grind metals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty