Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adenine sulfate dihydrate ≥97%

Supplier: Thermo Fisher Scientific
Warning
Synonyms: 9H-Purin-6-amine hemisulphate dihydrate6-Aminopurine sulphate dihydrate6-Aminopurine sulfate dihydrateAdenine hemisulfate dihydrateAdenine hemisulphate dihydrate7H-Purin-6-amine hemisulfate dihydrate9H-Purin-6-amine hemisulfate dihydrate7H-Purin-6-amine hemisulphate dihydrate

J10450-22
101170-670EA 254.22 USD InStock
101170-670
Adenine sulfate dihydrate ≥97%
Adenine sulphate dihydrate


Formula: (C₅H₅N₅)₂·H₂SO₄·2H₂O
MW: 404.37 g/mol
Melting Pt: ca 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00213655
CAS Number: 321-30-2
EINECS: 206-286-5
Merck Index: 14,00152
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty