Would you like to log in again?
SHIBAM Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
A146
76121-390PK 95.85 USD InStock
76121-390
SHIBAM Agar, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Used as a selective and differential growth medium for the cultivation of Shiga toxin-producing <italic>E.coli </italic>(STEC).
  • Detects enterohemolysin producing E. coli
  • Zones of clear blood hemolysis (beta hemolysis) surrounding the colonies are associated with Shiga-toxin producing E. coli
  • Formulation recommended by BAM
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty