Would you like to log in again?
Caffeic Acid Agar (Bird Seed Agar), Hardy Diagnostics
Supplier: Hardy Diagnostics
G213
76169-676PK 125.59 USD InStock
76169-676
Caffeic Acid Agar (Bird Seed Agar), Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the selective isolation and differentiation of Cryptococcus neoformans and Cryptococcus gattii
Also known as Bird Seed Agar or Niger Seed Agar.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty