Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Strep B Carrot Broth™ One-Step, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z40 Z44BX Z46BX
76121-438PK 707.04 USD InStock
76121-438 76121-436 76121-440
Strep B Carrot Broth™ One-Step, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the detection of hemolytic Group B Strep in pregnant women
  • Easy one-step process - place sample directly in the tube
  • Simple read-out - development of any orange to red color whatsoever, indicates a positive result
  • Accurate results - for positives, there is no need for further sub-culturing or testing
  • Low cost - lower cost when compared to the LIM broth method
  • Easy to use - can be used with liquid and gel-based transport systems
  • Multiple formats - carrot broth media is available in multiple formats that work on many automated inoculating machines, such as the WASP® or BD Innova®
Strep B Carrot Broth™ One-Step is a selective and differential medium which is intended for the detection of Group B Streptococcus (GBS) from anovaginal specimens collected from pregnant women. The medium is used as an aid in the qualitative determination of GBS colonization in pregnant women. The color change reaction from white to orange is representative of a positive result for presence of hemolytic GBS.
The one-step test is an improvement over conventional methods by increasing sensitivity, decreasing turn-around time, and lowering overall cost. All components are now included in the color change test, so the use of additional tiles is no longer necessary. Positives require no follow up testing.
The new tile-less system allows for fast results for the cultivation and identification of beta-hemolytic strains of Group B Streptococcus.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty