Would you like to log in again?
V9 Agar, for Mycology, Hardy Diagnostics
Supplier: Hardy Diagnostics
G98
76177-592PK 101.4 USD InStock
76177-592
V9 Agar, for Mycology, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
V9 Agar (Potato Flake Agar with V8 Juice) is recommended for the isolation, cultivation and rapid sporulation of fungi
  • Optically clear polystyrene - for distortion free viewing
  • Heavy-duty - reduces breakage during shipping
  • Stacking rings - polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
  • Breathable packaging - breathable bag that prevents buildup of condensation and excess moisture
The plates are deep-filled to reduce the effects of drying during prolonged incubation.
Packaging: Packaged in sleeves of 10 plates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty