Would you like to log in again?
CIN Agar/MacConkey Agar with Sorbitol, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
J49
76196-466PK 45.55 USD InStock
76196-466
CIN Agar/MacConkey Agar with Sorbitol, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
CIN Agar/MacConkey with Sorbitol used for the isolation of Yersinia, Aeromonas and E-coli O157. Incubate at 37 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty