Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bicine 98.5-101.5% (dry basis), ultrapure

Supplier: Thermo Fisher Scientific
Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acidN,N-Bis(2-hydroxyethyl)glycine

J12091-A1
101172-252EA 530.09 USD InStock
101172-252
Bicine 98.5-101.5% (dry basis), ultrapure
Bicine


Formula: C₆H₁₃NO₄
MW: 163.17 g/mol
Melting Pt: ca 191 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004295
CAS Number: 150-25-4
EINECS: 205-755-1
Merck Index: 14,01201
Order Now
Specification Test Results
Assay (Dry Basis) 98.5 - 101.5%
Assay Absorbance (250 nm, 10%, 1 cm) ≤0.1
Identification (FTIR) Conforms
Loss on Drying ≤0.5%
pKa (20 °C) 8.35 ±0.15
Iron (Fe) ≤30 ppm
Lead (Pb) ≤10 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty