Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Peptone for Bacteriology, Ultrapure, Thermo Scientific

Supplier: Thermo Scientific
J20048-Q1 J20048-P5
101169-642EA 1342.11 USD InStock
101169-642 101172-856
Peptone for Bacteriology, Ultrapure, Thermo Scientific
Media for Microbiology Dehydrated Media for Microbiology
An extremely high grade of peptone prepared specifically for general bacteriological purposes.

  • Form: Powder
  • Ash: ≤ 15%
  • Loss on Drying: ≤ 6%

Caution: Store at ambient temperatures.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty