Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrophenyl-ɑ-L-fucopyranoside

Supplier: Thermo Fisher Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:


J34131-03
101170-626EA 1330 USD InStock
101170-626
4-Nitrophenyl-ɑ-L-fucopyranoside
4-Nitrophenyl-alpha-L-fucopyranoside


Formula: C₁₂H₁₅NO₇
MW: 285.25 g/mol
Melting Pt: 194…198 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063697
CAS Number: 10231-84-2
EINECS: 233-553-3
Order Now
Specification Test Results
Identification (FTIR) Conforms

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty