Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mineral oil light for molecular biology

Supplier: Thermo Fisher Scientific

J71600-K2
101173-288EA 200.31 USD InStock
101173-288
Mineral oil light for molecular biology
Mineral oil light


Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5
EINECS: 232-455-8
Order Now
Specification Test Results
Assay Functional Assay Functionally tested for use in PCR
Assay Readily Carbonizable Substances Passes test
Neutrality Passes test
Identification (FTIR) Conforms
Impurity Content Limit of Polynuclear Compounds Passes test
Impurity Content Solid Paraffin Passes test
Specific Gravity (25 °C) 0.818 to 0.880
Viscosity (40 °C) ±33.5 centistokes

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty