Would you like to log in again?
Endothelin Converting Enzyme 1 Activity Assay Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB273323-100TEST
76200-958EA 867.51 USD InStock
76200-958
Endothelin Converting Enzyme 1 Activity Assay Kit (Fluorometric), BioVision, Inc.
Assays Enzyme Assays
Industry's first kit to detect ECE1 activity in biological samples
  • Simple one-step reaction
  • Takes only 1-2 hrs
  • Non-radiometric fluorescent detection
  • HTP adaptable
  • Detection of ECE1 activity in tissue/cell lysates
Endothelin Converting Enzyme 1 (ECE1; EC: 3.4.24.71) is a membrane-bound metalloprotease that cleaves inactive big endothelin (big ET-1), which is the precursor of active endothelin. Studies have shown that big ET-1 could be used as a predictor of survival in patients suffering esophageal squamous cell carcinoma. ECE-1 is abundantly expressed in vivo in endothelial cells producing mature ET-1. As of today, Endothelin is the most potent vasoconstrictor known and it has cytokine- or hormone-like activities. Therefore, ECE-1 could play a significant role in pathogenesis of cardiovascular diseases and Alzheimer’s Disease. BioVision’s Endothelin Converting Enzyme 1 Activity Assay Kit utilizes the ability of active ECE-1 to cleave a synthetic substrate (MCA-based peptide) releasing free fluorophore, in the presence or absence of the ECE-1 Inhibitor Mix. The released f.luorophore can be easily quantified using a fluorescence microplate reader. Our kit uses a unique combination of substrate and inhibitor that specifically detects ECE-1 in a variety of biological samples. Contribution of other enzymes with similar catalytic properties - i.e. Endothelin Converting Enzyme 2 (ECE-2), Angiotensin-Converting Enzyme (ACE1, ACE2), and Neprilysin is compensated for within the assay. Our assay kit is simple, specific and can detect as low as 0.5 µU of ECE-1 activity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty