Would you like to log in again?
Pyrophosphate (PPi) Assay Kit (Fluorometric/Colorimetric), BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB234040-100TEST
76200-962EA 736.62 USD InStock
76200-962
Pyrophosphate (PPi) Assay Kit (Fluorometric/Colorimetric), BioVision
Assays Enzyme Assays
Simple, Highly Sensitive, HTS adaptable assay to measure Homocysteine in biological samples.
  • Determination of PPi concentration in biological samples
  • Monitor pyrophosphate release by a variety of enzymes
  • Screening inhibition or activities of enzymes that consume pyrophosphate
  • Simple, sensitive and HTP adaptable Protocol
  • Specific for Pyrophosphate: other molecules containing phosphate groups do not interference with the assay
  • This kit can detect as little as 2 µM of Pyrophosphate in a variety of biological samples
Inorganic pyrophosphate (diphosphate, pyrophosphoric acid or PPi) is a small molecule formed during, or required as a substrate for, a number of biochemical reactions. Enzymatic reactions that generate PPi include ATP hydrolysis, DNA and RNA polymerization, cyclic AMP formation, and enzymatic activation of fatty acids to form their CoA esters. Conversely, enzymes that utilize PPi may include cyclases, hydrolases and ligases. Measurement of inorganic pyrophosphate (PPi) in plasma, serum and other biologic fluids is important in studies of bone metabolism, renal stone disease, and certain types of arthritis. BioVision’s Pyrophosphate (PPi) Assay Kit provides a fast, convenient and ultrasensitive method for determination of free inorganic pyrophosphate levels in biological material. PPi produced during biotic processes is detected through a series of reactions which utilize a proprietary enzyme mix and probe, generating a stable product that can be quantified by either colorimetric or fluorometric readout. Generated fluorescence (Ex/Em = 535/587 nm) or color (OD: 570 nm) intensities are directly proportional to the concentrations of pyrophosphate, enabling precise measurements. Monomeric inorganic phosphate (Pi) does not interfere with the assay. Our assay delivers an easy and robust method suitable for use in a variety of biological samples and can be performed in a convenient microtiter-plate format. The kit provides sufficient reagents for 100 fluorometric or 50 colorimetric assays, respectively. This kit can detect as low as 1.8 μM PPi in plasma and serum samples.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty