Would you like to log in again?
EZClick™ O-GlcNAc Modified Glycoprotein Assay Kit (FACS/Microscopy, Green Fluorescence), BioVision
Supplier: BioVision
AB235633-100TEST
76200-964EA 817.34 USD InStock
76200-964
EZClick™ O-GlcNAc Modified Glycoprotein Assay Kit (FACS/Microscopy, Green Fluorescence), BioVision
Assays Protein Assays
Highly specific method for labeling and detection of O-GlcNAc-glycosylated proteins within cells.
  • Simple, fast, does not require lengthy incubation times
  • Applications- This assay provides a convenient and accurate procedure to measure glycoproteins (O-GlcNAc Modified Proteins) in biological samples.
  • Detection method- Flow Cytometry (Ex/Em 488/(530/590) nm) and Fluorescence Microscopy
  • Robust method for labeling and detection of O-GlcNAc-glycosylated proteins within cells.
Glycans are vital components of glycoproteins, glycolipids, and proteoglycans in all domains of life. Glycoproteins are grouped by the type of carbohydrate and amino acid linkage site. N-linked glycosylation is a modification of asparagine, whereas O-linked glycosylation occurs through the hydroxyl group of serine and threonine residues. Glycosylation occurs co- or post-translationally on >50% of eukaryotic proteins resulting in membrane-associated, intracellular, or secreted glycoproteins that are crucial in cellular processes, protein bioactivity and metabolic turnover. Modification by O-linked-N-acetyl glucosamine (O-GlcNAc) has rapidly emerged as a major cellular signaling mechanism with number of modified targets similar to protein phosphorylation. Many oncogenes and tumor suppressors are regulated by O-GlcNAcylation. O-GlcNAc modification is ubiquitous among eukaryotes, from yeast to humans and its modifying enzymes have been well characterized. O-GlcNAc modified nuclear and cytosolic targets include: transcription factors, signaling proteins, metabolic enzymes, mitochondrial trafficking, cell cycle regulation, glucose homeostasis. O-GlcNAc glycosylation is implicated in normal brain functions, etiology of neurodegeneration, type II diabetes, and pathways involved in morphogenesis and virulence factors of microbes and plant host cells. Since glycoproteins are not directly encoded in the genome, methods of characterization and analyses of glycoproteins are of great interest. Thus BioVision offers EZClickTM O-GlcNAc Modified Glycoprotein Assay Kit, a highly specific, simple and robust method for labeling and detection of O-GlcNAc-glycosylated proteins within cells. We use a modified glucosamine precursor that is fed directly into the cells, processed by the hexosamine pathway and incorporated into the proteins. Followed by click reaction with alkyne-containing dye, this system offers a powerful method for imaging the localization, trafficking, and dynamics of glycans, or detection by FACS for quantitative studies. Labeled Glycoproteins can be directly detected in 1D or 2D gels using the appropriate excitation sources, or enriched by immunoprecipitation with biotin-alkyne or antibodies prior to proteomic analysis. We provide sufficient materials for 100 assays in a 96-well plate format
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty