Would you like to log in again?
Uricase Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB234042-100TEST
76200-966EA 821.19 USD InStock
76200-966
Uricase Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
Quick, sensitive & HTS adaptable assay for measurement of uricase activity in a variety of samples.
  • Simple procedure; takes ~ 1 hour
  • Fast and convenient
  • The assay is sensitive, stable and high-throughput adaptable.
  • Rapid assessment of Uricase activity in mammalian and plant tissues
Uricase (factor-independent urate hydroxylase, urate oxidase (UO); EC 1.7.3.3) is an enzyme involved in the uric acid metabolism. Uric acid is the end product of purine metabolism, and high levels of uric acid in blood causes gout. Uricase is present in a wide range of mammals but absent from human beings. It has been widely used for the measurement of uric acid concentrations in biological samples and recombinant uricase (rasburicase) has been used as a drug for the prevention of tumor lysis syndrome. BioVision’s Uricase Activity Assay Kit provides a quick and easy method for the measurement of uricase activity in a wide variety of samples. In this assay, uricase oxidizes uric acid forming a product that reacts with the probe, thus generating a fluorescence signal (Ex/Em= 535/587nm). The generated fluorescence signal is directly proportional to the amount of active uricase present in the sample. This kit provides a sensitive and high throughput adaptable assay and can measure uricase activity in biological samples as low as 10 µU.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty