Would you like to log in again?
Achromobacter lyticus Lysyl Endopeptidase
Supplier: Fujifilm Wako Chemicals USA
129-02541 125-02543
100369-826EA 719.23 USD InStock
100369-826 100369-822
Achromobacter lyticus Lysyl Endopeptidase
Enzymes
Lysyl Endopeptidase® is a proteolytic enzyme which is applied in medicine, food processing and as reagent in the synthesis and analysis of proteins.
  • High specificity and efficiency of protein digestion allow for easy database searches by peptide mass
  • Improved cleavage at lysine residue and increase in number of peptides obtained in combination with trypsin
  • In-gel digestion
  • Activity: indicated on label
  • Isoelectric point: 6.9 to 7.0
This enzyme which was originally isolated from Achromobacter lyticus M497-1 catalyzes the highly selective carboxy-terminal cleavage of the peptide bonds at both the lysine and S-aminoethylcysteine residues. It has a complete activity retention after an incubation of up to 6 hours in 4 M urea or 0.1% SDS solution at 30 °C.
Caution: For research use only.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty