Would you like to log in again?
BioMix™ Red DNA Polymerase Mix, Bioline Reagents
Supplier: Bioline
BioMix™
BIO-25006
490005-934EA 295.71 USD InStock
490005-934
BioMix™ Red DNA Polymerase Mix, Bioline Reagents
Nucleic Acid Reagents End-point PCR Enzymes and Kits
BioMix™ Red is a complete ready-to-use 2x reaction mix containing an ultra-stable Taq DNA polymerase
  • Versatile
  • High throughput and convenient
  • Direct gel loading
  • Suitable for routine PCR applications and TA cloning
It contains an additional inert red dye that permits easy visualization and direct loading onto a gel.
Delivery information: BioMix Red is supplied with additional MgCl2 solution and any fine adjustments be required.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty