Would you like to log in again?
Conductivity Calibration Pouches, Cole-Parmer
Supplier: Cole Parmer
35653-11
11025-102EA 79.91 USD InStock
11025-102
Conductivity Calibration Pouches, Cole-Parmer
Electrochemical Meters Conductivity Meters
Single-use 20 mL pouches are sealed to ensure freshness, take these convenient buffer solutions anywhere.
  • A µS-to-ppm conversion table
  • Printed on each pack for quick reference
Ordering information: Includes 20 single-use pouches in a dispensing box, with NIST-traceable calibration certificate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty