Would you like to log in again?
GranuCult™ Tryptic Soy agar (TSA), USP, EMD Millipore
Supplier: EMD Millipore
1.05458.5000
EM1.05458.5000EA 1467.79 USD InStock
EM1.05458.5000
GranuCult™ Tryptic Soy agar (TSA), USP, EMD Millipore
Media for Microbiology Dehydrated Media for Microbiology
  • Synonyms: Casein-peptone soyameal-peptone agar, Cacein-peptone soyameal-peptone agar
  • Stability test (colour and hemolysis): Non-hemolytic
Certifications: EP, USP, JP, ISO and FDA-BAM GranuCult™ certified.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty