Would you like to log in again?
ESF SFM Mammalian Cell Culture Medium, Expression Systems
Supplier: Expression Systems
98-001
103194-134EA 110.77 USD InStock
103194-134
ESF SFM Mammalian Cell Culture Medium, Expression Systems
Cell Culture Media
ESF SFM™ Mammalian Cell Culture Medium is an animal origin-free formulation and is developed for robust cell growth and protein expression for a wide range of mammalian cells.
  • Complete, ready-to-use, contains L-glutamine
  • Optimized for robust cell growth and protein expression
  • Available in 1L bottles
  • Appropriate for suspension and adherent cultures (contains surfactant)
  • Ideal for scale-up manufacturing
  • Inquire about custom formulations and packaging for bioreactors
  • Adapted cells available in suspension
ESF SFM cell culture medium is a serum-free, animal-free formulation designed for growth and expression of HEK 293, CHO and hybridoma cells in suspension. It is designed for use in shake flasks or bioreactors.
Delivery information: Standard Ground
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty