Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calbiochem® Superoxide Dismutase Assay Kit II, MilliporeSigma

Supplier: MilliporeSigma
Calbiochem®
574601-1KIT
EM574601-1KITEA 723.08 USD InStock
EM574601-1KIT
Calbiochem® Superoxide Dismutase Assay Kit II, MilliporeSigma
Assays Enzyme Assays
Superoxide dismutase assay kit II is a sensitive spectrophotometric assay (450 nm) for the measurement of all three types (Cu/Zn, Mn, and Fe) of SOD.

  • Positive Control: Bovine erythrocyte SOD (Cu/Zn)
  • The amount of enzyme needed to exhibit 50% dismutation of the superoxide radical

Delivery information: Kit contains assay buffer, sample buffer, radical detector, SOD standard, xanthine oxidase, 96-Well plate, plate cover, and a user protocol

Packaging: Packed in a glass bottle.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty