Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BioSim™ Bevacizumab (Avastin®) (Human) ELISA Kit, BioVision, Inc.

Supplier: BioVision
BioSim™
AB237642-96TEST
76199-276EA 2206.31 USD InStock
76199-276
BioSim™ Bevacizumab (Avastin®) (Human) ELISA Kit, BioVision, Inc.
Assays ELISAs
A Sandwich ELISA kit for the quantitative measurement of Bevacizumab in human serum or plasma.

  • Easy, convenient and time-saving method
  • Detection Range: 30 – 1000 ng/mL
  • Sensitivity: 30 ng/mL

Bevacizumab is a recombinant human IgG1 monoclonal antibody specific for all human vascular endothelial growth factor-A (VEGF-A) isoforms. The humanized anti-VEGF monoclonal antibody, bevacizumab, has been approved by the FDA as a first-line treatment for metastatic colorectal cancer in combination with chemotherapy. The pharmacokinetic properties of bevacizumab in several species have been previously described and are consistent with a typical humanized monoclonal antibody. It was shown in the literature that the surveillance of circulating concentration during maintenance therapy represents a direct and/or indirect factor for some other side effects. Identification of biomarkers for (non-)response and risk factors for adverse drug reactions that might be related to serum concentrations and maintaining the effective concentration of Bevacizumab in order to potentially avoid some side effects with a reliable method might be beneficial. BioVision’s BioSimTM Bevacizumab ELISA kit has been developed for specific quantification of Bevacizumab concentration in human serum or plasma with high sensitivity and reproducibility.

Cross Reactivity: No significant cross-reactivity or interference with other proteins present in native human serum or other therapeutic immunoglobulins.Assay Precision: Intra-Assay: CV <15%; Inter-Assay: CV <15% (CV (%) = SD/mean X 100), Recovery rate: 85 – 115% with normal human serum samples with known concentrations.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty