Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

QuickDetect™ Low Molecular Weight Heparin (LMWH) (Hu) ELISA Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

QuickDetect™
AB285268-96TEST
76199-688EA 1432.61 USD InStock
76199-688
QuickDetect™ Low Molecular Weight Heparin (LMWH) (Hu) ELISA Kit, BioVision
Assays ELISAs
A rapid sandwich ELISA kit for quantifying of LMWH in human serum and other biological fluid.

  • Easy, convenient and time-saving
  • Performed within 90 minutes
  • Detection Range: 3.7 – 120 ng/mL

Low-molecular-weight heparin (LMWH) is a class of anticoagulant medications. They are used in the prevention blood clots and treatment of venous thromboembolism (deep vein thrombosis and pulmonary embolism) and in the treatment of myocardial infarction. It has less unpredictable side effect than naturally derived heparin.

BioVision's QuickDetectTM LMWH ELISA kit is a sandwich ELISA assay for measuring the quantity of human LMWH in serum, plasma and cell culture and other biological fluid. The entire process can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results.

Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty