Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-5-Hydroxymethylcytosine Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
905-904-0050
76201-746EA 296.03 USD InStock
76201-746
Anti-5-Hydroxymethylcytosine Rabbit Polyclonal Antibody
Antibodies
Recommended Applications: ELISA, IP, Immunoblot

Species reactivity: Species independent

Type: Primary
Antigen: 5-Hydroxymethylcytosine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Species independent

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty