Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD15 Rabbit Monoclonal Antibody [clone: FUT4/1478R]

Supplier: Enzo Life Sciences
ABS358-0100
76201-804EA 600.38 USD InStock
76201-804
Anti-CD15 Rabbit Monoclonal Antibody [clone: FUT4/1478R]
Antibodies
Recombinant monoclonal antibody

  • Recommended Applications: Flow Cytometry, IF, IHC (PS)
  • Species reactivity: Human, Mouse, Rat

Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: FUT4/1478R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty