Would you like to log in again?
NanoBRET Positive Control, 2 × 20 µg, Promega
Supplier: Promega
N1581
PAN1581EA 337.49 USD InStock
PAN1581
NanoBRET Positive Control, 2 × 20 µg, Promega
Vectors, Plasmids and Libraries
The NanoBRET Positive Control Vector encodes a NanoLuc-HaloTag fusion protein that tethers together the NanoLuc donor and HaloTag acceptor proteins to ensure efficient energy transfer.
The NanoBRET Positive Control Vector encodes a NanoLuc-HaloTag fusion protein that tethers together the NanoLuc donor and HaloTag acceptor proteins to ensure efficient energy transfer. The NanoBRET Positive Control Vector should be diluted in Transfection Carrier DNA to reduce cellular expression levels due to the high NanoBRET signal produced. The control vector can be used when performing instrument setup or as a reference control when performing other NanoBRET assays. The NanoBRET Positive Control contains the following key features: 1) NanoLuc-HaloTag fusion gene. 2) Kanamycin resistance gene for selection in bacteria. 3) A CMV promoter for high-level expression of NanoLuc-HaloTag fusion in mammalian cells. The Transfection Carrier DNA contains the following key features: Ampicillin resistance gene for selection in bacteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty