Would you like to log in again?
NanoBRET PPI Control Pair (p53, MDM2), 2 × 20 µg, Promega
Supplier: Promega
N1641
PAN1641EA 687.84 USD InStock
PAN1641
NanoBRET PPI Control Pair (p53, MDM2), 2 × 20 µg, Promega
Vectors, Plasmids and Libraries
The NanoBRET PPI Control Pair (p53, MDM2) is an optimized pair of vectors designed to measure the interaction of protein partners p53 and MDM2 using the NanoBRET assay.
  • The NanoBRET PPI Control Pair (p53, MDM2) is an optimized pair of vectors designed to measure the interaction of protein partners p53 and MDM2 using the NanoBRET assay.
The NanoBRET PPI Control Pair (p53, MDM2) is an optimized pair of vectors designed to measure the interaction of protein partners p53 and MDM2 using the NanoBRET assay. The p53, MDM2 assay gives a strong NanoBRET signal and can be used when performing instrument setup or included as a reference control pair when performing other NanoBRET PPI assays. The p53-HaloTag Fusion Vector contains the following key features: 1) p53-HaloTag fusion gene. 2) Kanamycin resistance gene for selection in bacteria. 3) A CMV promoter for high-level expression of the p53-HaloTag fusion in mammalian cells. The NanoLuc-MDM2 Fusion Vector contains the following key features: 1) NanoLuc-MDM2 fusion gene. 2) Kanamycin resistance gene for selection in bacteria. 3) A CMV promoter for high-level expression of the NanoLuc-MDM2 fusion in mammalian cells.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty