Would you like to log in again?
pF4A CMV Flexi Vector, 20 µg, Promega
Supplier: Promega
C8481
PAC8481EA 402.89 USD InStock
PAC8481
pF4A CMV Flexi Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
Designed for high-level expression of proteins in mammalian cells from the CMV promoter without a selectable marker
  • Untagged Proteins Expressed in Mammalian Cells Without a Selectable Marker
  • Expresses high-levels of protein from cloned insert with CMV promoter
  • Transfer your cloned insert into other Flexi(R) Vectors for additional applications (e.g., bacterial or cell-free protein expression) as needed
  • Adapts to high-throughput formats for large screening projects, enhancing productivity
The Flexi Vector System is a simple, yet powerful, directional cloning method for protein-coding sequences. It is based on two rare-cutting restriction enzymes, SgfI and PmeI, and provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence. The pF4A and pF4K CMV Flexi Vectors below are designed specifically for high-level expression of proteins in mammalian cells from the CMV promoter without a selectable marker. All inserts may be confirmed by cell-free expression with the TNT T7 Quick System (Cat.# L1170). Note: Flexi Vectors contain the lethal barnase gene to reduce background colonies without inserts during the subcloning procedure. Using the Flexi Vector Cloning System replaces the barnase gene with your insert. These vectors, as purchased, cannot be cultured in normal laboratory strains of E. coli without an insert.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty