Would you like to log in again?
CheckMate Positive Control Vectors, 1 set, Promega
Supplier: Promega
CheckMate™
C9370
PAC9370EA 290.31 USD InStock
PAC9370
CheckMate Positive Control Vectors, 1 set, Promega
Vectors, Plasmids and Libraries
Powerful system for detecting protein:protein interactions in vivo.
  • Powerful System for Detecting Protein:Protein Interactions In Vivo
  • Mammalian system for studying interactions in cell line of choice
  • Study post-translational modifications including glycosylation, phosphorylation and acylation
Two-hybrid systems are extremely powerful methods for detecting protein:protein interactions in vivo. The basis of two-hybrid systems is the modular domains found in some transcription factors: a DNA-binding domain, which binds to a specific DNA sequence, and a transcriptional activation domain, which interacts with the basal transcriptional machinery. A transcriptional activation domain in association with a DNA-binding domain will promote the assembly of RNA polymerase II complexes at the TATA box and increase transcription. In the CheckMate Mammalian Two-Hybrid System the DNA-binding domain and the transcriptional activation domain, produced by separate plasmids, are closely associated when one protein (X) fused to a DNA-binding domain interacts with a second protein (Y) fused to a transcriptional activation domain. In this system, interaction between proteins X and Y results in transcription of a reporter gene.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty