Would you like to log in again?
pBR322 Vector, 10 µg, Promega
Supplier: Promega
D1511
PAD1511EA 82.08 USD InStock
PAD1511
pBR322 Vector, 10 µg, Promega
Vectors, Plasmids and Libraries
pBR322 Vector.
  • Plasmid Subcloning Vector that Carries Genes for Tetracycline and Ampicillin Resistance
  • Supplied at a concentration of 1mg/ml
The plasmid pBR322 Vector carries the genes for tetracycline and ampicillin resistance. pBR322 DNA digests typically are used as molecular weight size markers in gel analysis of nucleic acids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty