Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disposable Polypropylene Test Tubes (8×50mm), 1000/pack, Promega

Supplier: Promega
E4221
PAE4221EA 275.48 USD InStock
PAE4221
Disposable Polypropylene Test Tubes (8×50mm), 1000/pack, Promega
Tubes Microcentrifuge Tubes
These accessories are available for the now-discontinued 20/20 luminometer.

  • For Use With TD-20/20 Instrument
  • Accessories only for discontinued Turner BioSystems Luminometer Model TD-20/20
  • Not for Turner BioSystems 20/20 n Single-Tube Luminometer or Promega GloMax(R) 20/20 Luminometer
  • Power Supply now discontinued

These accessories are only for the now-discontinued Turner BioSystems Luminometer Model TD-20/20. They are not for the Turner BioSystems 20/20 n Single-Tube Luminometer or Promega GloMax 20/20 Luminometer. Turner BioSystems and Promega have teamed up to develop the next-generation luminometers for single-tube and 96-well plate analysis. For more information, see the GloMax Luminometer catalog pages or contact Technical Support.

Storage: 15 to 30 °C

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty