Would you like to log in again?
EZCut™ HRV14 3C (Precission) Protease Agarose Beads, BioVision Inc.
Supplier: BioVision
AB286883-25ML AB286883-5ML AB286883-1ML
76208-682EA 6197.63 USD InStock
76208-682 76209-564 76209-562
EZCut™ HRV14 3C (Precission) Protease Agarose Beads, BioVision Inc.
Protein Purification Protein Purification Kits
Prepared by covalent coupling active HRV14 3C Protease to activated 4% cross-linked agarose beads.
  • It is highly selective and designed for efficient cleavage of recombinant fusion proteins
  • Very specific in recognization
  • Easy and convenient procedure
Active HRV14 3C Protease (Precission Protease) domain from human rhinovirus type 14 has high selectivity and site-specificity against Precission cleavage site (Leu-Glu-Val-Leu-Phe-Gln/Gly-Pro), thereby allowing for the production of recombinant proteins with minimal number of extra amino acids. BioVision’s EZCut™ HRV14 3C (Precission) Protease Agarose is prepared by covalent coupling active HRV14 3C Protease to activated 4% cross-linked agarose beads. EZCut™ HRV14 3C (Precission) Protease Agarose Beads are designed for efficient cleavage of recombinant fusion proteins, containing the Precission protease cleavage site, circumventing the need for chromatographic techniques to remove the protease after cleavage reaction is completed. The optimal temperature for cleavage is 30°C; however, the enzyme is active over wide ranges of temperature. These beads can be regenerated for repeat use and the reusability depends on the usage and handling of the product.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty