Would you like to log in again?
biOrb® TUBE Aquaria
Supplier: GEOGLOBAL PARTNERS
45969 45939 72073 72071 45944 72064 72072 45979 72062 72070 45984 45934 72063 45929 45974 72065
470234-830EA 234 USD InStock
470234-830 470336-916 470336-914 470336-918 470336-908 470234-832 470336-920 470234-822 470234-834 470234-824 470336-912 470234-836 470234-826 470336-910 470234-828 470336-922
biOrb® TUBE Aquaria
Animal Supplies Animal Environmental Enrichment
A Full 360 Degree View!
  • Five stage filtration
  • Easy feeding access
  • Platform base
The biOrb® TUBE Aquarium offers an unobstructed, 360 degree view of its underwater world. This exquisite 4 or 9 gallon aquarium is designed with cylindrical contouring and is available in sleek black or white trim.
The biOrb® TUBE is equipped with a standard white LED with an easy on and off switch. The biOrb® TUBE with LED light uses the same proven filtration system as the rest of the biOrb® range. Like all biOrbs, the biOrb® TUBE uses ceramic media to provide biological filtration which has a surface area equivalent to the size of a soccer field. Any excess waste is collected and held in a filter cartridge at the base of the biOrb®. To maintain your aquarium all you do is simply replace the filter cartridge every four weeks and change a third of the water. Also for your peace of mind, the light and air pump are low voltage. The aquarium itself is made from acrylic which is ten times stronger than glass and backed by a 2 year warranty.
Each TUBE includes the tank, a lid, the light, an air pump and air stone, ceramic media, water conditioner, and a filter cartridge. You just supply the water and fish!
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty